missing translation for 'onlineSavingsMsg'
Learn More

Flutamide, MedChemExpress

Product Code. 16480758
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480758 5 g 5g
16422039 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16480758 Supplier Medchem Express Supplier No. HYB0022
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Flutamide is an antiandrogen drug, with its active metablolite binding at androgen receptor with Ki values of 55 nM, and primarily used to treat prostate cancer.

Flutamide is an antiandrogen drug, with its active metablolite binding at androgen receptor with Ki values of 55 nM, and primarily used to treat prostate cancer.

Specifications

Chemical Name or Material Flutamide
Physical Form Solid
CAS 13311-84-7
Color Yellow
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C11H11F3N2O3
For Use With (Application) Cancer-programmed cell death
Synonym SCH 13521
Solubility Information DMSO : 100 mg/mL (362.04 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.36 mM; Need ultrasonic)
SMILES CC(C)C(NC1=CC=C([N+]([O-])=O)C(C(F)(F)F)=C1)=O
Molecular Weight (g/mol) 276.21
Formula Weight 276.21
Percent Purity 99.53%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.