missing translation for 'onlineSavingsMsg'
Learn More

Flusilazole, TRC

Product Code. 30396982
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396982 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396982 Supplier Toronto Research Chemicals Supplier No. TRCF59880025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flusilazole
CAS 85509-19-9
Molecular Formula C16 H15 F2 N3 Si
Quantity 25 g
Synonym 1-[[Bis(4-fluorophenyl)methylsilyl]methyl]-1H-1,2,4-triazole, Benocarp, Bis(4-fluorophenyl)methyl(1H-1,2,4-triazole-1-ylmethyl)silane, Cane Strike, Capitan, Cursor, DPD 78710F, DPX-H 6573, DPX-N 6573, Flusilazol, Flusilazole, Genie, Genie (fungicide), Genie 25, Lyric, Nustar, Olymp, Olymp 10EX, Punch, Punch (pesticide), Punch 40EC, Punch Bis, Sanction
InChI Formula InChI=1S/C16H15F2N3Si/c1-22(12-21-11-19-10-20-21,15-6-2-13(17)3-7-15)16-8-4-14(18)5-9-16/h2-11H,12H2,1H3
SMILES C[Si](Cn1cncn1)(c2ccc(F)cc2)c3ccc(F)cc3
IUPAC Name bis(4-fluorophenyl)-methyl-(1,2,4-triazol-1-ylmethyl)silane
Molecular Weight (g/mol) 315.39
Formula Weight 315.1003
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.