missing translation for 'onlineSavingsMsg'
Learn More

Fluroxypyr, TRC

Product Code. 30390107
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390107 100 mg 100mg
30392452 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390107 Supplier Toronto Research Chemicals Supplier No. TRCF598780100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluroxypyr
CAS 69377-81-7
Purity Grade Notes HPLC
Molecular Formula C7 H5 Cl2 F N2 O3
Quantity 100 mg
Synonym Acetic acid, [(4-amino-3,5-dichloro-6-fluoro-2-pyridinyl)oxy]- (9CI), 2-[(4-Amino-3,5-dichloro-6-fluoro-2-pyridinyl)oxy]acetic acid, EF 689, Fluroxypyr, Shuangzhiling, Starane Advanced, [(4-Amino-3,5-dichloro-6-fluoro-2-pyridyl)oxy]acetic acid
InChI Formula InChI=1S/C7H5Cl2FN2O3/c8-3-5(11)4(9)7(12-6(3)10)15-1-2(13)14/h1H2,(H2,11,12)(H,13,14)
SMILES Nc1c(Cl)c(F)nc(OCC(=O)O)c1Cl
IUPAC Name 2-(4-amino-3,5-dichloro-6-fluoropyridin-2-yl)oxyacetic acid
Molecular Weight (g/mol) 255.03
Formula Weight 253.9661
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.