missing translation for 'onlineSavingsMsg'
Learn More

Flurofamide-d4, TRC

Product Code. 30400340
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400340 10 mg 10mg
30408602 1 mg 1mg
30402220 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30400340 Supplier Toronto Research Chemicals Supplier No. TRCF59668710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flurofamide-d4
Molecular Formula C7 D4 H5 F N3 O2 P
Quantity 10 mg
Synonym 4-Fluoro-N-(diaminophosphinyl)benzamide-d4, Flurfamide-d4, NSC 342995-d4
InChI Formula InChI=1S/C7H9FN3O2P/c8-6-3-1-5(2-4-6)7(12)11-14(9,10)13/h1-4H,(H5,9,10,11,12,13)/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c(C(=O)NP(=O)(N)N)c([2H])c([2H])c1F
IUPAC Name 2,3,5,6-tetradeuterio-N-diaminophosphoryl-4-fluorobenzamide
Molecular Weight (g/mol) 221.162
Formula Weight 221.067
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.