missing translation for 'onlineSavingsMsg'
Learn More

Fluridone, TRC

Product Code. 30396328
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396328 100 mg
30400933 250 mg
30399970 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396328 Supplier Toronto Research Chemicals Supplier No. TRCF598770100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59756-60-4
Molecular Formula C19 H14 F3 N O
Molecular Weight (g/mol) 329.32
Synonym Fluridone, 4(1H)-Pyridinone, 1-methyl-3-phenyl-5-[3-(trifluoromethyl)phenyl]- (9CI, ACI), 1-Methyl-3-phenyl-5-[3-(trifluoromethyl)phenyl]-4(1H)-pyridinone (ACI), EL 171, Fluridone, Sonar
IUPAC Name 1-methyl-3-phenyl-5-[3-(trifluoromethyl)phenyl]pyridin-4-one
SMILES CN1C=C(C(=O)C(=C1)c2cccc(c2)C(F)(F)F)c3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H14F3NO/c1-23-11-16(13-6-3-2-4-7-13)18(24)17(12-23)14-8-5-9-15(10-14)19(20,21)22/h2-12H,1H3
Formula Weight 329.1027
Chemical Name or Material Fluridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.