missing translation for 'onlineSavingsMsg'
Learn More

Flurbiprofen Glyceryl Ester, TRC

Product Code. 30403239
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403239 50 mg
30408464 10 mg
30398192 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403239 Supplier Toronto Research Chemicals Supplier No. TRCG61522850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124635-84-3
Molecular Formula C18H19FO4
Molecular Weight (g/mol) 318.34
Synonym 2-Fluoro-α-methyl-[1,1'-Biphenyl]-4-acetic Acid 2,3-Dihydroxypropyl Ester, 2,3-Dihydroxypropyl 2-(2-fluoro-[1,1'-biphenyl]-4-yl)propanoate
SMILES CC(C1=CC=C(C2=CC=CC=C2)C(F)=C1)C(OCC(O)CO)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H19FO4/c1-12(18(22)23-11-15(21)10-20)14-7-8-16(17(19)9-14)13-5-3-2-4-6-13/h2-9,12,15,20-21H,10-11H2,1H3
Formula Weight 318.13
Chemical Name or Material Flurbiprofen Glyceryl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.