missing translation for 'onlineSavingsMsg'
Learn More

Flurbiprofen Acyl-Beta-D-glucuronide (Mixture of Diastereomers), TRC

Product Code. 30389090
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389090 1 mg
30390899 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389090 Supplier Toronto Research Chemicals Supplier No. TRCF5987151MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91683-37-3
Molecular Formula C21 H21 F O8
Molecular Weight (g/mol) 420.38
Synonym 1-(2-Fluoro-α-methyl[1,1'-biphenyl]-4-acetate)-β-D-glucopyranuronic Acid, Flurbiprofen Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-6-[2-(3-fluoro-4-phenyl-phenyl)propanoyloxy]-3,4,5-trihydroxy-tetrahydropyran-2-carboxylic acid
SMILES CC(C(=O)O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O)c2ccc(c(F)c2)c3ccccc3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H21FO8/c1-10(12-7-8-13(14(22)9-12)11-5-3-2-4-6-11)20(28)30-21-17(25)15(23)16(24)18(29-21)19(26)27/h2-10,15-18,21,23-25H,1H3,(H,26,27)/t10?,15-,16-,17+,18-,21-/m0/s1
Formula Weight 420.122
Chemical Name or Material Flurbiprofen acyl-beta-D-Glucuronide (mixture of diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.