missing translation for 'onlineSavingsMsg'
Learn More

Flurandrenolide, TRC

Product Code. 30393483
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393483 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393483 Supplier Toronto Research Chemicals Supplier No. TRCF5986502.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flurandrenolide
CAS 1524-88-5
Molecular Formula C24 H33 F O6
Quantity 2.5 mg
Synonym Pregn-4-ene-3,20-dione, 6α-fluoro-11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone (7CI,8CI), (6α,11β,16α)-6-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregn-4-ene-3,20-dione, 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregn-4-ene-3,20-dione deriv., 33379, 6α-Fluoro-11β,21-dihydroxy-16α,17α-(isopropylidenedioxy)pregn-4-ene-3,20-dione, 6α-Fluoro-16α-hydroxyhydrocortisone 16,17-acetonide, Alondra F, Cordran, Drenison, Drocort, Fluadrenolone, Fludroxicortide, Fludroxycortide, Fluorandrenolone, Fluorandrenolone acetonide, Fluoroandrenolone acetonide, Flurandrenolide, Flurandrenolone, Flurandrenolone acetonide, Haelan, Haldrone F, L 33379, Sermaka
InChI Formula InChI=1S/C24H33FO6/c1-21(2)30-19-9-14-13-8-16(25)15-7-12(27)5-6-22(15,3)20(13)17(28)10-23(14,4)24(19,31-21)18(29)11-26/h7,13-14,16-17,19-20,26,28H,5-6,8-11H2,1-4H3/t13-,14-,16-,17-,19+,20+,22-,23-,24+/m0/s1
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)CC[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
IUPAC Name (6α,11β,16α)-6-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregn-4-ene-3,20-dione
Molecular Weight (g/mol) 436.51
Formula Weight 436.2261
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.