missing translation for 'onlineSavingsMsg'
Learn More

Flurandrenolide, MedChemExpress

Product Code. 16450376
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16450376

Brand: Medchem Express HYB1013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fludroxycortide is a synthetic topical steroid and is used as an anti-inflammatory treatment for use on skin irritations.

Fludroxycortide is a synthetic topical steroid and is used as an anti-inflammatory treatment for use on skin irritations.

Specifications

Chemical Name or Material Flurandrenolide
CAS 1524-88-5
Color White
Purity Grade Notes Research
Molecular Formula C24H33FO6
Quantity 2 mg
Synonym Fludroxycortide Flurandrenolone
Solubility Information DMSO : 100 mg/mL (229.09 mM; Need ultrasonic)
SMILES C[C@@]12[C@@]3(C(CO)=O)[C@@](OC(C)(O3)C)([H])C[C@@]1([H])[C@]4([H])C[C@H](F)C5=CC(CC[C@]5(C)[C@@]4([H])[C@@H](O)C2)=O
Molecular Weight (g/mol) 436.51
Formula Weight 436.51
Percent Purity 98.68%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.