missing translation for 'onlineSavingsMsg'
Learn More

Flupirtine Maleate, MedChemExpress

Product Code. p-200096337
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16447258

Brand: Medchem Express HY17001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Flupirtine Maleate is a brain penetrant, and orally bioavailable, non-opioid and centrally acting analgesic agent. Flupirtine Maleate is an indirect N-methyl-D-aspartate receptor (NMDAR) antagonist. Neuroprotective properties.

Flupirtine Maleate is a brain penetrant, and orally bioavailable, non-opioid and centrally acting analgesic agent. Flupirtine Maleate is an indirect N-methyl-D-aspartate receptor (NMDAR) antagonist. Neuroprotective properties.

Specifications

Chemical Name or Material Flupirtine Maleate
CAS 75507-68-5
Color White
Purity Grade Notes Research
Molecular Formula C19H21FN4O6
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (237.87 mM; Need ultrasonic) ∣H2O : 1.43 mg/mL (3.40 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NC1=CC=C(N=C1N)NCC2=CC=C(C=C2)F)OCC.O=C(/C=C\C(O)=O)O
Molecular Weight (g/mol) 420.39
Formula Weight 420.39
Percent Purity 98.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.