missing translation for 'onlineSavingsMsg'
Learn More

Flupirtine Maleate, MedChemExpress

Product Code. 16447258
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447258 10 mg 10mg
16407226 100 mg 100mg
16403277 50 mg 50mg
16401826 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16447258 Supplier Medchem Express Supplier No. HY17001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Flupirtine Maleate is a brain penetrant, and orally bioavailable, non-opioid and centrally acting analgesic agent. Flupirtine Maleate is an indirect N-methyl-D-aspartate receptor (NMDAR) antagonist. Neuroprotective properties.

Flupirtine Maleate is a brain penetrant, and orally bioavailable, non-opioid and centrally acting analgesic agent. Flupirtine Maleate is an indirect N-methyl-D-aspartate receptor (NMDAR) antagonist. Neuroprotective properties.

Specifications

Chemical Name or Material Flupirtine Maleate
CAS 75507-68-5
Color White
Purity Grade Notes Research
Molecular Formula C19H21FN4O6
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (237.87 mM; Need ultrasonic) ∣H2O : 1.43 mg/mL (3.40 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NC1=CC=C(N=C1N)NCC2=CC=C(C=C2)F)OCC.O=C(/C=C\C(O)=O)O
Molecular Weight (g/mol) 420.39
Formula Weight 420.39
Percent Purity 98.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.