missing translation for 'onlineSavingsMsg'
Learn More

Fluphenazine, TRC

Product Code. 30135425
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135425 50 mg 50mg
30003522 10 mg 10mg
30006983 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135425 Supplier Toronto Research Chemicals Supplier No. TRCF59841850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-23-8
Molecular Formula C22H26N3OSF3 (.) di-HCl
Molecular Weight (g/mol) 510.44
Synonym 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-, 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]- (6CI,7CI,8CI), 4-[3-[2-(Trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-1-piperazineethanol, 1-(2-Hydroxyethyl)-4-[3-(trifluoromethyl-10-phenothiazinyl)propyl]piperazine, 10-[3-(2-Hydroxyethyl)piperazinopropyl]-2-(trifluoromethyl)phenothiazine, 4-[3-(2-Trifluoromethyl-10-phenothiazyl)-propyl]-1-piperazineethanol, 4-[3-[2-(Trifluoromethyl)phenothiazin-10-yl]propyl]-1-piperazine ethanol, Elinol, Fluorfenazine, Fluorophenazine, Fluorphenazine, Fluphenazine, Ftorphenazine, Pacinol, Phthorphenazine, S 94, SQ 4918, Siqualine, Siqualon, Triflumethazine, Vespazine
IUPAC Name 2-[4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol
SMILES OCCN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H26F3N3OS/c23-22(24,25)17-6-7-21-19(16-17)28(18-4-1-2-5-20(18)30-21)9-3-8-26-10-12-27(13-11-26)14-15-29/h1-2,4-7,16,29H,3,8-15H2
Formula Weight 437.1749
Percent Purity >95%
Chemical Name or Material Fluphenazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.