missing translation for 'onlineSavingsMsg'
Learn More

Fluphenazine Sulfoxide (Fluphenazine Decanoate EP Impurity A), TRC

Product Code. 30396981
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396981 5 mg
30390492 10 mg
30402107 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396981 Supplier Toronto Research Chemicals Supplier No. TRCF5984155MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1674-76-6
Molecular Formula C22 H26 F3 N3 O2 S
Molecular Weight (g/mol) 453.52
Synonym 1-Piperazineethanol, 4-[3-[5-oxido-2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-, 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-, S-oxide, 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]-, S-oxide (7CI,8CI), 4-[3-[5-Oxido-2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-1-piperazineethanol, Fluphenazine S-oxide, Fluphenazine sulfoxide, 2-[4-[3-[5-Oxo-2-(trifluoromethyl)-10H-5lambda4-phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol, Fluphenazine Dihydrochloride Imp. A (EP)
IUPAC Name 2-[4-[3-[5-oxo-2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol
SMILES OCCN1CCN(CCCN2c3ccccc3S(=O)c4ccc(cc24)C(F)(F)F)CC1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H26F3N3O2S/c23-22(24,25)17-6-7-21-19(16-17)28(18-4-1-2-5-20(18)31(21)30)9-3-8-26-10-12-27(13-11-26)14-15-29/h1-2,4-7,16,29H,3,8-15H2
Formula Weight 453.1698
Percent Purity >95%
Chemical Name or Material Fluphenazine S-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.