missing translation for 'onlineSavingsMsg'
Learn More

Fluphenazine N4-Oxide Sulphoxide, TRC

Product Code. 30397575
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397575 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397575 Supplier Toronto Research Chemicals Supplier No. TRCF59834510MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluphenazine N4-Oxide Sulfoxide
CAS 76005-61-3
Molecular Formula C22 H26 F3 N3 O3 S
Quantity 10 mg
Synonym 4-[3-[5-Oxido-2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-1-piperazineethanol 1-Oxide, 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-, S,1-dioxide, Fluphenazine Sulfoxide N1-Oxide
InChI Formula InChI=1S/C22H26F3N3O3S/c23-22(24,25)17-6-7-21-19(16-17)27(18-4-1-2-5-20(18)32(21)31)9-3-8-26-10-12-28(30,13-11-26)14-15-29/h1-2,4-7,16,29H,3,8-15H2
SMILES OCC[N+]1([O-])CCN(CCCN2c3ccccc3S(=O)c4ccc(cc24)C(F)(F)F)CC1
IUPAC Name 2-[1-oxido-4-[3-[5-oxo-2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-ium-1-yl]ethanol
Molecular Weight (g/mol) 469.52
Formula Weight 469.1647
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.