missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluphenazine Dihydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Fluphenazine dihydrochloride |
| CAS | 146-56-5 |
| Molecular Formula | C22 H26 F3 N3 O S . 2 Cl H |
| Quantity | 5 g |
| Synonym | Fluphenazine Dihydrochloride, 4-[3-[2-(Trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-1-piperazineethanol hydrochloride (1:2), 4-[3-[2-(Trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-1-piperazineethanol dihydrochloride, 1-(2-Hydroxyethyl)-4-[3-(2-trifluoromethyl-10-phenothiazinylpropyl]piperazine dihydrochloride, 10-[3-[4-(2-Hydroxyethyl)piperazinyl]propyl]-2-trifluoromethylphenothiazine dihydrochloride, 4-[3-[2-(Trifluoromethyl)phenothiazin-10-yl]propyl]-1-piperazineethanol dihydrochloride, Anatensol, Dapotum, Fludecan, Flufenazin, Fluorophenazine dihydrochloride, Fluphenazine chloride, Fluphenazine dihydrochloride, Fluphenazine hydrochloride, Liogen, Lyogen, Lyorodin, Mirenil, Moditen, OMCA, Permitil, Permitil hydrochloride, Prolinate, Prolixin, Siqualone, Squibb 4918, Tensofin, Trancin, Valamina |
| InChI Formula | InChI=1S/C22H26F3N3OS.2ClH/c23-22(24,25)17-6-7-21-19(16-17)28(18-4-1-2-5-20(18)30-21)9-3-8-26-10-12-27(13-11-26)14-15-29;;/h1-2,4-7,16,29H,3,8-15H2;2*1H |
| SMILES | Cl.Cl.OCCN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC Name | 2-[4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol;dihydrochloride |
| Molecular Weight (g/mol) | 510.44 |
| Formula Weight | 509.1282 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?