missing translation for 'onlineSavingsMsg'
Learn More

Fluphenazine Beta-D-Glucuronide, TRC

Product Code. 30402770
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402770 1 mg 1mg
30389660 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Produktkod. 30402770 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF5984101MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Fluphenazine beta-D-Glucuronide
CAS 133310-09-5
Molecular Formula C28 H34 F3 N3 O7 S
Quantity 1 mg
InChI Formula InChI=1S/C28H34F3N3O7S/c29-28(30,31)17-6-7-21-19(16-17)34(18-4-1-2-5-20(18)42-21)9-3-8-32-10-12-33(13-11-32)14-15-40-27-24(37)22(35)23(36)25(41-27)26(38)39/h1-2,4-7,16,22-25,27,35-37H,3,8-15H2,(H,38,39)/t22-,23-,24+,25-,27+/m0/s1
SMILES O[C@@H]1[C@@H](O)[C@H](OCCN2CCN(CCCN3c4ccccc4Sc5ccc(cc35)C(F)(F)F)CC2)O[C@@H]([C@H]1O)C(=O)O
IUPAC Name (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[2-[4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethoxy]oxane-2-carboxylic acid
Molecular Weight (g/mol) 613.65
Formula Weight 613.207
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.