missing translation for 'onlineSavingsMsg'
Learn More

Flupentixol dihydrochloride, Thermo Scientific Chemicals

Product Code. 16594284 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16594284 5 g 5g
16504294 250 mg 250mg
16584284 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16594284 Supplier Thermo Scientific Acros Supplier No. 464820050
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2413-38-9
Molecular Weight (g/mol) 507.44

Specifications

Molecular Formula C23H25F3N2OS·2HCl
Quantity 5 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: insoluble in ethyl acetate, methanol
Formula Weight 507.44
Chemical Name or Material Flupentixol dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.