missing translation for 'onlineSavingsMsg'
Learn More

Flupentixol dihydrochloride, Thermo Scientific Chemicals

Product Code. 16504294 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16504294 250 mg 250mg
16584284 1 g 1g
16594284 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16504294 Supplier Thermo Scientific Acros Supplier No. 464822500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2413-38-9
Molecular Weight (g/mol) 507.44
InChI Key IOVDQEIIMOZNNA-MHKBYHAFSA-N
IUPAC Name dihydrogen 2-(4-{3-[(9Z)-2-(trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl}piperazin-1-yl)ethan-1-ol dichloride
SMILES [H+].[H+].[Cl-].[Cl-].OCCN1CCN(CC\C=C2\C3=CC=CC=C3SC3=CC=C(C=C23)C(F)(F)F)CC1

Specifications

Molecular Formula C23H27Cl2F3N2OS
Quantity 250 mg
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: insoluble in ethyl acetate, methanol
Formula Weight 507.44
Chemical Name or Material Flupentixol dihydrochloride
compliance-icons
Product Identifier
  • Flupentixol dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Flupentixol dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.