missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Flupenthixol dihydrochloride

Product Code. 18732664
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18732664 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18732664 Supplier R&D Systems™ Supplier No. 4057/50
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Dopamine receptor antagonist

Antipsychotic drug. Exhibits antagonistic activity at dopamine D1-5 receptors. Used in schizophrenia treatment.

This compound is a mixture of E/Z isomers (ratio: 60:40).

TRUSTED_SUSTAINABILITY

Specifications

CAS 2413-38-9
Quantity 50 mg
Product Line Tocris
Molecular Formula C23H27Cl2F3N2OS
Formula Weight 507.44
Purity >99%
Molecular Weight (g/mol) 507.437
Chemical Name or Material Flupenthixol dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.