missing translation for 'onlineSavingsMsg'
Learn More

Fluoxymesterone, 97+%

Product Code. 10513481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10513481 1 g 1g
11986881 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
Product Code. 10513481 Supplier Thermo Scientific Acros Supplier No. 124590010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.

Fluoxymesterone, CAS # 76-43-7, is a potent, orally administered, synthetic steroid with androgenic activity. It is used in the treatment of male hypogonadism. | CAS: 76-43-7 | C20H29FO3 | 336.447 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Fluoxymesterone is a white, odorless, crystalline powder, soluble in pyridine
  • It is an orally-administered synthetic steroid that is about 5-10 times more potent than male steroid, testosterone
  • It is metabolized in the liver, and less than 5% of this drug is excreted from the body unchanged
  • Adverse effects of fluoxymesterone include sodium and water retention leading to edema, acne, cardiovascular toxicity, and hepatotoxicity

Applications

  • Fluoxymesterone has been found to competitively inhibit prolactin or estrogen receptors in tumorigenic breast cells, and is therefore administered in the treatment of breast cancer
  • In males, this compound is used as a replacement in conditions associated with symptoms of deficiency or absence of endogenous testosterone
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-43-7
Molecular Formula C20H29FO3
Molecular Weight (g/mol) 336.447
InChI Key YLRFCQOZQXIBAB-RBZZARIASA-N
Synonym fluoxymesterone, fluoximesterone, fluoxymestrone, halotestin, androfluorene, androfluorone, fluotestin, androsterolo, fluosterone, flusteron
PubChem CID 6446
ChEBI CHEBI:5120
IUPAC Name (8S,9R,10S,11S,13S,14S,17S)-9-fluoro-11,17-dihydroxy-10,13,17-trimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
SMILES CC12CCC(=O)C=C1CCC3C2(C(CC4(C3CCC4(C)O)C)O)F

Specifications

Melting Point 240.0°C
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition + 104.00 (20.00°C c=1,EtOH)
Specific Rotation + 104.00
Solubility Information Solubility in water: negligible. Other solubilities: soluble in puridine, slightly soluble in acetone, chloroform, sparingly soluble in methanol, practically insoluble in ether, benzene, hexanes
Formula Weight 336.45
Chemical Name or Material Fluoxymesterone
compliance-icons
Product Identifier
  • Fluoxymesterone
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.