missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoxymesterone, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Fluoxymesterone
Specifications
Specifications
| Chemical Name or Material | Fluoxymesterone |
| CAS | 76-43-7 |
| Analyte or Component Names | Fluoxymesterone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H29 F O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Fluoxymesterone |
| InChI Formula | InChI=1S/C20H29FO3/c1-17-8-6-13(22)10-12(17)4-5-15-14-7-9-19(3,24)18(14,2)11-16(23)20(15,17)21/h10,14-16,23-24H,4-9,11H2,1-3H3/t14-,15-,16-,17-,18-,19-,20-/m0/s1 |
| SMILES | C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C |
| IUPAC Name | (8S,9R,10S,11S,13S,14S,17S)-9-fluoro-11,17-dihydroxy-10,13,17-trimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction