missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Fluoxetine (Hydrochloride)

Product Code. 17110182
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17110182 500 mg 500mg
17100182 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17110182 Supplier STEMCELL Technologies Supplier No. 73144

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Serotonin reuptake inhibitor

  • Fluoxetine is a selective serotonin reuptake inhibitor, displaying a distinct preference for the serotonin transporter (Kd = 0.81 nM) over both the norepinephrine and dopamine transporters (Kd values of 240 and 3600 nM, respectively; Tatsumi et al.) This product is supplied as a hydrochloride salt of the molecule
  • Induces proliferation of human embryonic stem cell-derived neural precursors, and enhances their neuronal differentiation (Chang et al. 2010)
  • Induces proliferation and inhibits differentiation of human hypothalamic neuroprogenitor cells (Sousa-Ferreira et al.)
  • Stimulates proliferation of mouse fetal neural stem cells (Chang et al. 2012)
  • Stimulates in vivo proliferation of amplifying neural progenitor cells in adult mouse brain (Encinas et al.)

Specifications

CAS 56296-78-7
Synonym Fontex™; LY110140; Prozac™; Sarafem™
Quantity 500 mg
Molecular Formula C17H18F3NO · HCl
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.