missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Fluoxetine (Hydrochloride)
Description
- Fluoxetine is a selective serotonin reuptake inhibitor, displaying a distinct preference for the serotonin transporter (Kd = 0.81 nM) over both the norepinephrine and dopamine transporters (Kd values of 240 and 3600 nM, respectively; Tatsumi et al.) This product is supplied as a hydrochloride salt of the molecule
- Induces proliferation of human embryonic stem cell-derived neural precursors, and enhances their neuronal differentiation (Chang et al. 2010)
- Induces proliferation and inhibits differentiation of human hypothalamic neuroprogenitor cells (Sousa-Ferreira et al.)
- Stimulates proliferation of mouse fetal neural stem cells (Chang et al. 2012)
- Stimulates in vivo proliferation of amplifying neural progenitor cells in adult mouse brain (Encinas et al.)
Specifications
Specifications
| CAS | 56296-78-7 |
| Synonym | LY110140 |
| Quantity | 100 mg |
| Molecular Formula | C17H18F3NO · HCl |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?