missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine.HCl, Lipomed

Product Code. 30460188
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460188 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460188 Supplier Lipomed Supplier No. LPMFLX721HC50
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Fluoxetine.HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluoxetine Hydrochloride
CAS 56296-78-7
Analyte or Component Names Fluoxetine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H18 F3 N O . Cl H
Synonym Fluoxetine Hydrochloride, Adofen, Affectine, Alzac 20, Ansilan, Deproxin, Digassim, Erocap, Fluctin, Fluctine, Fludac, Flufran, Flunil, Fluox-Puren, Fluoxac, Fluoxeren, Fluoxetine hydrochloride, Fluoxil, Flutin, Flutine, Fluxen, Fluxetyl, Fluxil, Fontex, Foxetin, LY 110140, Lilly 110140, Lorien, Lovan, Margrilan, Modipran, N-Methyl-3-(4-trifluoromethylphenoxy)-3-phenylpropylamine hydrochloride, Neupax, Nodepe, Nopres, Nuzak, Octozac, Oxedep, Pluzac, Pragmaten, Prizma, Proctin, Prodep, Profluzac, Prozac, Prozac 20, Reconcile, Reneuron, Rowexetina, Salipax, Sanzur, Sarafem, Sinzac, Zactin, Zedprex, Zepax, N-Methyl-gamma-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride (1:1), (+/-)-N-Methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propylamine hydrochloride
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CNCCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2
IUPAC Name N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 345.79
Quantity 50 mg
Formula Weight 345.1107 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.