missing translation for 'onlineSavingsMsg'
Learn More

(±)-Fluoxetine-d5 HCl (phenyl-d5), CDN

Product Code. 30324781
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Product Code. Quantity unitSize
30324781 0.01 g 0.01g
30313343 0.005 g 0.01g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30324781

Brand: CDN CDND57490.01G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluoxetine-d5 Hydrochloride
CAS 1173020-43-3
Molecular Formula C17 2H5 H13 F3 N O . Cl H
Quantity 0.01 g
Synonym N-Methyl-3-(2,3,4,5,6-pentadeuteriophenyl)-3-[4-(trifluoromethyl)phenoxy]propan-1-amine hydrochloride, Fluoxetine-D5 Hydrochloride
InChI Formula InChI=1 S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21 H,11-12H2,1H3;1 H/i2D,3 D,4 D,5 D,6 D;
SMILES Cl.[2 H]c1c([2 H])c([2 H])c(C(CCNC)Oc2ccc(cc2)C(F)(F)F)c([2 H])c1[2 H]
IUPAC Name N-methyl-3-(2,3,4,5,6-pentadeuteriophenyl)-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
Molecular Weight (g/mol) 350.82
Formula Weight 350.1421 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.