missing translation for 'onlineSavingsMsg'
Learn More

Fluorosilicic Acid, 35%, Technical, Fisher Chemical™

Product Code. 10519000 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
HDPE plastic bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10519000 500 mL HDPE plastic bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10519000 Supplier Fisher Chemical Supplier No. F/1400/08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 16961-83-4
Physical Form Liquid
Quantity 500 mL
Molecular Formula F6H2Si
MDL Number 36289
Synonym hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure
InChI Key OHORFAFFMDIQRR-UHFFFAOYSA-P
SMILES [H+].[H+].F[Si-2](F)(F)(F)(F)F
IUPAC Name hexafluorosilicon(2-);hydron
Molecular Weight (g/mol) 144.091
PubChem CID 21863527
Formula Weight 144.09g/mol
Packaging HDPE plastic bottle
Show More Show Less
compliance-icons
Product Identifier
  • Fluorosilicic acid, 35%
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Fluorosilicic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.