missing translation for 'onlineSavingsMsg'
Learn More

Fluoronitrofen, TRC

Product Code. 30393970
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393970 250 mg
30395348 100 mg
30407599 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393970 Supplier Toronto Research Chemicals Supplier No. TRCF156430250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13738-63-1
Molecular Formula C12H6Cl2FNO3
Molecular Weight (g/mol) 302.09
Synonym Ether, 2,4-dichloro-6-fluorophenyl p-nitrophenyl (8CI), 1,5-Dichloro-3-fluoro-2-(4-nitrophenoxy)benzene, 2,4-Dichloro-6-fluoro-4'-nitrodiphenyl ether, 2,4-Dichloro-6-fluorophenyl 4-nitrophenyl ether, 2,4-Dichloro-6-fluorophenyl-p-nitrophenyl ether, 2-Fluoro-4,6-dichloro-4'-nitrodiphenyl ether, 2-Fluoro-4,6-dichlorophenyl 4-nitrophenyl ether, 4-Nitro-2',4'-dichloro-6'-fluorodiphenyl ether, CFNP, Fluoronitrofen
IUPAC Name 1,5-dichloro-3-fluoro-2-(4-nitrophenoxy)benzene
SMILES [O-][N+](=O)c1ccc(Oc2c(F)cc(Cl)cc2Cl)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H6Cl2FNO3/c13-7-5-10(14)12(11(15)6-7)19-9-3-1-8(2-4-9)16(17)18/h1-6H
Formula Weight 300.9709
Chemical Name or Material Fluoronitrofen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.