missing translation for 'onlineSavingsMsg'
Learn More

Fluorometholone D Homo Derivative, TRC

Product Code. 30401578
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401578 1 mg 1mg
30393005 10 mg 10mg
30393006 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401578 Supplier Toronto Research Chemicals Supplier No. TRCF5979431MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorometholone D Homo Derivative
Analyte or Component Names Fluorometholone D Homo Derivative
Shipping Condition Room Temperature
Molecular Formula C22 H29 F O4
InChI Formula InChI=1S/C22H29FO4/c1-12-9-16-14-5-6-17(25)21(4,27)20(14,3)11-18(26)22(16,23)19(2)8-7-13(24)10-15(12)19/h7-8,10,12,14,16,18,26-27H,5-6,9,11H2,1-4H3/t12-,14-,16-,18-,19-,20-,21?,22-/m0/s1
Solution Type Neat
SMILES C[C@H]1C[C@H]2[C@@H]3CCC(=O)C(C)(O)[C@@]3(C)C[C@H](O)[C@]2(F)[C@@]4(C)C=CC(=O)C=C14
IUPAC Name (4aS,4bS,6S,10aS,10bR,11S,12aS)-10b-fluoro-1,11-dihydroxy-1,6,10a,12a-tetramethyl-3,4,4a,4b,5,6,11,12-octahydrochrysene-2,8-dione
Industry Type Pharmaceutical
Molecular Weight (g/mol) 376.46
Quantity 1 mg
Formula Weight 376.205
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.