missing translation for 'onlineSavingsMsg'
Learn More

Fluorometholone, 97%, Thermo Scientific Chemicals

Product Code. 15519513 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15519513 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15519513 Supplier Thermo Scientific Acros Supplier No. 460200010
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluorometholone, CAS # 426-13-1, is a glucocorticoid with anti-inflammatory properties.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Fluorometholone is a synthetic glucocorticoid
  • It has anti-inflammatory properties

Applications

  • Fluorometholone interacts with cytoplasmic glucocorticoid receptors resulting in activation of these receptors, which mediates gene expression
TRUSTED_SUSTAINABILITY

Specifications

CAS 426-13-1
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C22H29FO4
Quantity 1 g
Synonym fluorometholone, oxylone, flumetholon, fluormetholone, fml liquifilm, fluor-op, cortilet, delmeson, trilcin, fml forte
Solubility Information (2.5% in pyridine) clear colorless to light yellow
InChI Key FAOZLTXFLGPHNG-KNAQIMQKSA-N
SMILES CC1CC2C3CCC(C3(CC(C2(C4(C1=CC(=O)C=C4)C)F)O)C)(C(=O)C)O
IUPAC Name (6S,8S,9R,10S,11S,13S,14S,17R)-17-acetyl-9-fluoro-11,17-dihydroxy-6,10,13-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 376.46
PubChem CID 9878
ChEBI CHEBI:31625
Formula Weight 376.46
Percent Purity 97%
Chemical Name or Material Fluorometholone
Show More Show Less
compliance-icons
Product Identifier
  • Fluorometholone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.