missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorometholone 11,17-Diacetate, TRC
Specifications
Specifications
| Chemical Name or Material | Fluorometholone 11,17-Diacetate |
| Analyte or Component Names | Fluorometholone 11,17-Diacetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H33 F O6 |
| Synonym | 9-Fluoro-11beta,17-dihydroxy-6alpha-methylpregna-1,4-diene-3,20-dione Diacetate |
| InChI Formula | InChI=1S/C26H33FO6/c1-14-11-21-19-8-10-25(15(2)28,33-17(4)30)24(19,6)13-22(32-16(3)29)26(21,27)23(5)9-7-18(31)12-20(14)23/h7,9,12,14,19,21-22H,8,10-11,13H2,1-6H3/t14-,19-,21-,22-,23-,24-,25-,26-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@H]1C[C@H]2[C@@H]3CC[C@](OC(=O)C)(C(=O)C)[C@@]3(C)C[C@H](OC(=O)C)[C@]2(F)[C@@]4(C)C=CC(=O)C=C14 |
| IUPAC Name | [(6S,8S,9R,10S,11S,13S,14S,17R)-17-acetyl-17-acetyloxy-9-fluoro-6,10,13-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-11-yl] acetate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?