missing translation for 'onlineSavingsMsg'
Learn More

Fluorometholone 11,17-Diacetate, TRC

Product Code. 30389144
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389144 10 mg 10mg
30401237 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389144 Supplier Toronto Research Chemicals Supplier No. TRCF59316010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorometholone 11,17-Diacetate
Analyte or Component Names Fluorometholone 11,17-Diacetate
Shipping Condition Room Temperature
Molecular Formula C26 H33 F O6
Synonym 9-Fluoro-11beta,17-dihydroxy-6alpha-methylpregna-1,4-diene-3,20-dione Diacetate
InChI Formula InChI=1S/C26H33FO6/c1-14-11-21-19-8-10-25(15(2)28,33-17(4)30)24(19,6)13-22(32-16(3)29)26(21,27)23(5)9-7-18(31)12-20(14)23/h7,9,12,14,19,21-22H,8,10-11,13H2,1-6H3/t14-,19-,21-,22-,23-,24-,25-,26-/m0/s1
Solution Type Neat
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](OC(=O)C)(C(=O)C)[C@@]3(C)C[C@H](OC(=O)C)[C@]2(F)[C@@]4(C)C=CC(=O)C=C14
IUPAC Name [(6S,8S,9R,10S,11S,13S,14S,17R)-17-acetyl-17-acetyloxy-9-fluoro-6,10,13-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-11-yl] acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 460.535
Quantity 10 mg
Formula Weight 460.226
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.