missing translation for 'onlineSavingsMsg'
Learn More

Fluorol Yellow 088, TRC

Product Code. 30395160
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395160 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395160 Supplier Toronto Research Chemicals Supplier No. TRCF6217251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorol Yellow 088
CAS 81-37-8
Molecular Formula C22H16O
Quantity 1 g
Synonym Fluorol 5G, 2,8-Dimethylnaphtho[3,2,1-kl]xanthene, 2,8-Dimethylceroxene, C.I. 45550, C.I. Fluorescent Brightener 74, C.I. Fluorescent Brightening Agent 74, C.I. Solvent Green 4, Fluorescent Brightener 74, Fluorescent Green Dye, Fluorescent Green LF, Solvent Green 4
InChI Formula InChI=1S/C22H16O/c1-13-7-9-19-18(11-13)21-16-6-4-3-5-15(16)12-17-14(2)8-10-20(23-19)22(17)21/h3-12H,1-2H3
SMILES CN([C@@H]1[C@@H](N2CCCC2)C[C@]3(CCCO3)CC1)C(CC4=CC(Cl)=C(Cl)C=C4)=O
Molecular Weight (g/mol) 296.36
Formula Weight 296.1201
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.