missing translation for 'onlineSavingsMsg'
Learn More

Fluoroiodomethane, TRC

Product Code. 30400982
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400982 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400982 Supplier Toronto Research Chemicals Supplier No. TRCF5919555G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluoroiodomethane
CAS 373-53-5
Molecular Formula C H2 F I
Quantity 5 g
Synonym Fluoromethyl iodide
InChI Formula InChI=1S/CH2FI/c2-1-3/h1H2
SMILES FCI
IUPAC Name fluoro(iodo)methane
Molecular Weight (g/mol) 159.93
Formula Weight 159.9185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.