missing translation for 'onlineSavingsMsg'
Learn More

Fluoroimide, TRC

Product Code. 30006440
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006440 100 mg 100mg
30004230 1 g 1g
30005215 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006440 Supplier Toronto Research Chemicals Supplier No. TRCF591953100MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41205-21-4
Molecular Formula C10 H4 Cl2 F N O2
Molecular Weight (g/mol) 260.05
Synonym 3,4-Dichloro-1-(4-fluorophenyl)-1H-pyrrole-2,5-dione, 2,3-Dichloro-N-(4-fluorophenyl)maleimide, 2,3-Dichloro-N-(p-fluorophenyl)maleimide, Fluoroimide, Fluoromide, MK 23, N-(4-Fluorophenyl)-2,3-dichloromaleimide, N-(4-Fluorophenyl)dichloromaleimide, N-(p-Fluorophenyl)-2,3-dichloromaleimide, NSC 622384, Spartcide, Spat
IUPAC Name 3,4-dichloro-1-(4-fluorophenyl)pyrrole-2,5-dione
SMILES Fc1ccc(cc1)N2C(=O)C(=C(Cl)C2=O)Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H4Cl2FNO2/c11-7-8(12)10(16)14(9(7)15)6-3-1-5(13)2-4-6/h1-4H
Formula Weight 258.9603
Chemical Name or Material Fluoroimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.