missing translation for 'onlineSavingsMsg'
Learn More

Fluorocyclohexane, 97%

Product Code. 15530624 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15530624 5 g 5g
15540624 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15530624 Supplier Thermo Scientific Acros Supplier No. 459320050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 372-46-3 | C6H11F | 102.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-46-3
Molecular Formula C6H11F
Molecular Weight (g/mol) 102.15
InChI Key GOBGVVAHHOUMDK-UHFFFAOYSA-N
Synonym cyclohexane, fluoro, cyclohexyl fluoride, cyclohexylfluoride, fluoro-cyclohexane, acmc-1cpcv
PubChem CID 78988
IUPAC Name fluorocyclohexane
SMILES C1CCC(CC1)F

Specifications

Melting Point 13.0°C
Density 0.9280g/mL
Boiling Point 103.0°C to 105.0°C
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula C6H11F
Quantity 5 g
Specific Gravity 0.928
Formula Weight 102.15
Percent Purity 97%
Chemical Name or Material Fluorocyclohexane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Fluorocyclohexane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.