missing translation for 'onlineSavingsMsg'
Learn More

Fluoroboric acid, 50 wt% solution in water

Product Code. p-134477 Shop All Thermo Scientific Chemicals Products
Klik for at se tilgængelige muligheder
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Pakningsstørrelse:
25g
500g
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

Artikelnummer. 10236540

Brand: Thermo Scientific Acros 196700250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

CAS: 16872-11-0 | BF4H | 87.812 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16872-11-0, 7732-18-5, 10043-35-3
Molecular Formula BF4H
Molecular Weight (g/mol) 87.812
MDL Number MFCD00011345
InChI Key ODGCEQLVLXJUCC-UHFFFAOYSA-O
Synonym fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq
PubChem CID 28118
ChEBI CHEBI:38902
IUPAC Name hydron;tetrafluoroborate
SMILES [H+].[B-](F)(F)(F)F

Specifications

Color White
Melting Point -90.0°C
Boiling Point 130.0°C
Density 1.4100g/mL
Packaging Plastic bottle
Specific Gravity 1.41
Quantity 25 g
Linear Formula HBF4
Physical Form Adhering Crystals and/or Chunks
UN Number 1775
Fieser 01, 394
Merck Index 15, 4180
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol
Formula Weight 87.81
Percent Purity 48 to 51% (Acid)
Chemical Name or Material Fluoroboric acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Fluoroboric acid, 50 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.