missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ FluoroBlok™ 24 Well Permeable Supports and Companion Plates

Block light transmission, simplifying detection of migrating fluorescently labeled cells. Falcon™ FlouroBlok™ 24 Well Plate Permeable Supports are designed with a unique light-tight PET membrane. Must be used with Falcon™ 24 Well Companion Plate.

Brand:  Falcon™ 351151

Code : 66

Additional Details : Weight : 2.72160kg

 View more versions of this product

Product Code. 10375911

  • £430.00 / Pack of 48
Estimated Shipment: 25-09-2024
to see stock.
Add to basket

Description

Description

Falcon Cell Culture Insert Companion Plates have been specially designed for use with Falcon or Corning™ BioCoatTM Cell Culture Inserts so evaporation and contamination due to improper lid fit is eliminated.

In the Feeding Position, pipet access is improved for fluid handling on the basolateral side. Reagents can be added quickly and consistently for timed experiments. Aspiration of media from the well is easier, reducing the risk of contamination.

In the Incubation Position, Falcon Cell Culture Inserts remain locked in position in their Falcon Companion Plate wells. Media will not wick up between the insert and well wall. The unique Falcon low-evaporation lid provides a tortuous air passage system that reduces evaporation and contamination.

Specifications

Specifications

Cell Culture Insert, 3 μm Pore Size
PET
Cell Culture
24
48/Cs.
3 μm
0.65 cm
0.3 cm2
Sterile
Multi-Well Insert System
Falcon™ 24 Well TC-Treated Cell Polystyrene Permeable Support Companion Plate
1/Pk.
Polyethylene Terephthalate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers