missing translation for 'onlineSavingsMsg'
Learn More

Fluorobenzo[c]fluorene, TRC

Product Code. 30398007
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398007 100 mg 100mg
30398514 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398007 Supplier Toronto Research Chemicals Supplier No. TRCF588435100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorobenzo[c]fluorene
Analyte or Component Names Fluorobenzo[c]fluorene
Shipping Condition Room Temperature
Molecular Formula C17 H11 F
InChI Formula InChI=1S/C17H11F/c18-16-10-12-9-11-5-1-2-6-13(11)17(12)15-8-4-3-7-14(15)16/h1-8,10H,9H2
Solution Type Neat
SMILES Fc1cc2Cc3ccccc3c2c4ccccc14
IUPAC Name 5-fluoro-7H-benzo[c]fluorene
Industry Type Pharmaceutical
Molecular Weight (g/mol) 234.268
Quantity 100 mg
Formula Weight 234.084
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.