missing translation for 'onlineSavingsMsg'
Learn More

Fluorobenzene-d5, TRC

Product Code. 30395207
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395207 250 mg
30408265 100 mg
30403737 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30395207 Supplier Toronto Research Chemicals Supplier No. TRCF588302250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1423-10-5
Molecular Formula C6 2H5 F
Molecular Weight (g/mol) 101.13
Synonym Fluorobenzene-d5, Fluorobenzene D5
IUPAC Name 1,2,3,4,5-pentadeuterio-6-fluorobenzene
SMILES [2H]c1c([2H])c([2H])c(F)c([2H])c1[2H]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C6H5F/c7-6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D
Formula Weight 101.0689
Chemical Name or Material Fluorobenzene-D5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.