missing translation for 'onlineSavingsMsg'
Learn More

Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 99%

Product Code. 13555000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13555000 25 g Glass bottle 25g
10637400 1 g Glass bottle 1g
10361670 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13555000 Supplier Thermo Scientific Acros Supplier No. 434960250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 164298-23-1 | C5H12FN2·F6P | 264.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164298-23-1
Molecular Formula C5H12FN2·F6P
Molecular Weight (g/mol) 264.13
MDL Number MFCD02684443
InChI Key ZAVXOOLKAGPJPI-UHFFFAOYSA-N
Synonym tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, c5h12fn2.f6p, acmc-209dqu, tetramethyl fluoroformamidium hexafluorophosphate
PubChem CID 2774761
IUPAC Name [dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 107°C to 110°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 264.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate
compliance-icons
Product Identifier
  • Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.