missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | 104°C to 111°C |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 264.12 |
Percent Purity | 98% |
Chemical Name or Material | Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'