Learn More
WTW™ Fluoride Test Kits
Product Code. 11711083
Click to view available options
Range:
0.1 to 1.5 mg/L
Unit Size:
Pack of 25
Product Code. 11711083
Brand: WTW™ 252094
Specifications
For Use With (Application) For photometric analysis
For Use With (Equipment) Photometers
Includes Certificates
Range 0.1 to 1.5 mg/L
Test Method Photometric
Type Fluoride cell test kit
Product Content Correction
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Sekretesspolicy.
Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.