missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard Solution 1000ppm (1mg/mL), for Ion Pair Chromatography, ready to use, Fisher Chemical™

Product Code. 12486400 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12486400 500 mL 500mL
10568040 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12486400 Supplier Fisher Chemical Supplier No. J/4548/08
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluoride Standard Solution 1000ppm (1mg/mL), for Ion Pair Chromatography, ready to use

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 144315

Specifications

Packaging HDPE plastic bottle
Vapor Pressure 14mm Hg at 20°C deg C
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.