missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Fluoride Qualitative Test Paper

Product Code. 10117171 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color Indicator:
Pink to Yellow/White
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Indicator unitSize
10117171 Pink to Yellow/White Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10117171 Supplier Macherey-Nagel™ Supplier No. 90750
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Allows the rapid detection of fluoride ions in solutions containing hydrochlorid acid. Macherey-Nagel™ Fluoride Qualitative Test Paper is suitable for the safe and easy detection of dangerous hydrofluoric acid, which is used for example in computer chip production.

  • Simply add a drop of the hydrochlorid acid solution (pH ≤ 1) to the test paper and read immediately
  • High concentrations of fluorides are indicated by the appearance of a yellowish-white spot
  • Lesser amounts of flourides are indicated by a yellowish-white ring
  • Complex-bound fluoride ions react in the same manner as free fluoride ions
  • Limit of sensitivity: 20mg/L (ppm)
  • Interferences:
    • Chlorates and bromates; interference is eliminated by adding sodium dithionite (Na2S2O4)
    • Sulfate in very large amounts; interference can be eliminated by adding barium chloride

Specifications

Color Indicator Pink to Yellow/White
Format Strip
For Use With (Application) Determination of Fluoride
Parameter Specifications Limit of sensitivity: 20mg/L F
Quantity 200 Pack
Test Time Immediate
Type Qualitative Test Paper
Dimensions (L x W) 20 x 70 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.