missing translation for 'onlineSavingsMsg'
Learn More

Fluorescent Brightener 87 (Technical Grade), TRC

Product Code. 30397023
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397023 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397023 Supplier Toronto Research Chemicals Supplier No. TRCF40132010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorescent Brightener 87 (Technical Grade)
CAS 12768-91-1
Molecular Formula C40H40N12Na4O16S4
Quantity 10 g
Synonym Kayaphor S
InChI Formula InChI=1S/C40H44N12O16S4.4Na/c53-19-15-51(16-20-54)39-47-35(41-27-7-11-31(12-8-27)69(57,58)59)45-37(49-39)43-29-5-3-25(33(23-29)71(63,64)65)1-2-26-4-6-30(24-34(26)72(66,67)68)44-38-46-36(48-40(50-38)52(17-21-55)18-22-56)42-28-9-13-32(14-10-28)70(60,61)62;;;;/h1-14,23-24,53-56H,15-22H2,(H,57,58,59)(H,60,61,62)(H,63,64,65)(H,66,67,68)(H2,41,43,45,47,49)(H2,42,44,46,48,50);;;;/q;4*+1/p-4/b2-1+;;;;
SMILES OCCN(CCO)C1=NC(NC2=CC=C(S(=O)(O[Na])=O)C=C2)=NC(NC(C=C3S(=O)(O[Na])=O)=CC=C3/C=C/C(C=C4)=C(S(=O)(O[Na])=O)C=C4NC5=NC(N(CCO)CCO)=NC(NC6=CC=C(S(=O)(O[Na])=O)C=C6)=N5)=N1
IUPAC Name sodium (E)-6,6'-(ethene-1,2-diyl)bis(3-((4-(bis(2-hydroxyethyl)amino)-6-((4-sulfonatophenyl)amino)-1,3,5-triazin-2-yl)amino)benzenesulfonate)
Molecular Weight (g/mol) 1165.04
Formula Weight 1164.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.