missing translation for 'onlineSavingsMsg'
Learn More

Fluorescent Brightener 71 (~65%), TRC

Product Code. 30391041
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391041 2.5 g 2.5g
30406694 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391041 Supplier Toronto Research Chemicals Supplier No. TRCF5960802.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16090-02-1
Molecular Formula C40 H38 N12 O8 S2 . 2 Na
Molecular Weight (g/mol) 924.9149
Synonym 2,2'-Stilbenedisulfonic acid, 4,4'-bis[(4-anilino-6-morpholino-s-triazin-2-yl)amino]-, disodium salt (6CI,7CI,8CI), Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[[4-(4-morpholinyl)-6-(phenylamino)-1,3,5-triazin-2-yl]amino]-, disodium salt (9CI), 2M/G-LT, 4,4'-Bis(2-morpholino-4-anilino-s-triazinyl-6-amino)stilbene-2,2'-disulfonic acidisulfonic acid d disodium salt, 4,4'-Bis[(4-anilino-6-morpholino-1,3,5-triazin-2-yl)amino]stilbene-2,2'-disulfonic acid disodium salt, 4,4'-Bis[(6-anilino-4-morpholino-1,3,5-triazin-2-yl)amino]-2,2'-disulfostilbene disodium salt, 4,4'-Bis[4''-anilino-6''-morpholinotriazinyl-2''-amino]stilbene-2,2'-disulfonic acid disodium salt, Belofor KD 2, Belotex KD, Blancophor BBH, Blankophor BBH, Blankophor DML, Blankophor MBBH, Brightener 33, Brightener BR 15, C.I. 40625, C.I. Fluorescent Brightener 244, C.I. Fluorescent Brightener 250, C.I. Fluorescent Brightener 260, C.I. Fluorescent Brightener 71, CXT, Calcofluor White RC, D-MS, DAS 1, DAS 1 (brightener), DMA-X, DMS, Disodium 4,4'-bis(2-anilino-4-morpholino-1,3,5-triazin-6-ylamino)-2,2'-stilbenedisulfonate, Disodium 4,4'-bis(2-morpholino-4-phenylamino-s-triazin-6-ylamino)stilbine-2,2'-disulfonate, Disodium 4,4'-bis(4-anilino-2-morpholino-s-triazin-6-ylamino)stilbene-2,2'-disulfonate, Disodium 4,4'-bis(4-anilino-6-morpholino-s-triazin-2-ylamino)stilbene-2,2'-disulfonate, Disodium 4,4'-bis[(2-anilino-4-morpholino-s-triazin-6-yl)amino]-2,2'-stilbenedisulfonate, Disodium 4,4'-bis[(4-anilino-6-morpholino-1,3,5-triazin-2-yl)amino]stilbene-2,2'-disulfonate, Disodium anilinomorpholinotriazinyl-aminostilbenesulfonate, FB 71, Fluorescent Brightener 260, Fluorescent Brightener 71, Fluorescent Brightener CXT, Heliofor 3BC, Heliofor BDC, Hiltamine Arctic White DML, Kayaphor MAS, Leukopur PAM, MBBH 766, Mikephor TB, Optiblanc 2M/G-LT, Optiblanc 2MG, Phorwite MBBH 766, Photine CBUS, Photine CBUS 560B, Photine CBUS-B 560, Rylux D, Sodium 4,4'-bis(2-anilino-4-morpholino-6-amino-1,3,5-triazinyl)stilbene-2,2'-disulfonate, Stralex MD, Tinopal AMS, Tinopal AMS-GX, Tinopal DMA, Tinopal DMA-X, Tinopal DMS, Tinopal DMS-X, Tinopal EMS, Whitex SA, Whitex SA extra conc.(H)
IUPAC Name disodium;5-[(4-anilino-6-morpholin-4-yl-1,3,5-triazin-2-yl)amino]-2-[(E)-2-[4-[(4-anilino-6-morpholin-4-yl-1,3,5-triazin-2-yl)amino]-2-sulfonatophenyl]ethenyl]benzenesulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)c1cc(Nc2nc(Nc3ccccc3)nc(n2)N4CCOCC4)ccc1\C=C\c5ccc(Nc6nc(Nc7ccccc7)nc(n6)N8CCOCC8)cc5S(=O)(=O)[O-]

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C40H40N12O8S2.2Na/c53-61(54,55)33-25-31(43-37-45-35(41-29-7-3-1-4-8-29)47-39(49-37)51-17-21-59-22-18-51)15-13-27(33)11-12-28-14-16-32(26-34(28)62(56,57)58)44-38-46-36(42-30-9-5-2-6-10-30)48-40(50-38)52-19-23-60-24-20-52;;/h1-16,25-26H,17-24H2,(H,53,54,55)(H,56,57,58)(H2,41,43,45,47,49)(H2,42,44,46,48,50);;/q;2*+1/p-2/b12-11+;;
Formula Weight 924.2172
Chemical Name or Material Fluorescent Brightener 71
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.