missing translation for 'onlineSavingsMsg'
Learn More

Fluorescent Brightener 28 (Technical Grade), TRC

Product Code. 30134154
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134154 250 mg 250mg
30137141 1 g 1g
30003745 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134154 Supplier Toronto Research Chemicals Supplier No. TRCF599740250MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4404-43-7
Molecular Formula C40H44N12O10S2
Molecular Weight (g/mol) 916.98
Synonym C.I. Fluorescent Brightening Agent 28, 2,2'-(1,2-Ethenediyl)bis[5-[[4-[bis(2-hydroxyethyl)amino]-6-(phenylamino)-1,3,5-triazin-2-yl]amino]benzenesulfonic acid, 4,4'-Bis[(4-anilino-6-[bis(2-hydroxyethyl)amino]-1,3,5-triazin-2-yl)amino]stilbene-2,2'-disulfonic acid, 4,4'-Bis[2-anilino-4-[bis(hydroxyethyl)amino]-s-triazin-6-ylamino]-2,2'-stilbenedisulfonic acid, 4,4'-Bis[[4-anilino-6-bis[(2-hydroxyethyl)amino]-s-triazin-2-yl]amino]-2,2'-stilbenedisulfonic acid, Phorwite BBH
SMILES OCCN(CCO)c1nc(Nc2ccccc2)nc(Nc3ccc(\C=C\c4ccc(Nc5nc(Nc6ccccc6)nc(n5)N(CCO)CCO)cc4S(=O)(=O)O)c(c3)S(=O)(=O)O)n1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C40H44N12O10S2/c53-21-17-51(18-22-54)39-47-35(41-29-7-3-1-4-8-29)45-37(49-39)43-31-15-13-27(33(25-31)63(57,58)59)11-12-28-14-16-32(26-34(28)64(60,61)62)44-38-46-36(42-30-9-5-2-6-10-30)48-40(50-38)52(19-23-55)20-24-56/h1-16,25-26,53-56H,17-24H2,(H,57,58,59)(H,60,61,62)(H2,41,43,45,47,49)(H2,42,44,46,48,50)/b12-11+
Chemical Name or Material Fluorescent Brightener 28 (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.