Learn More
Description
Fluoresces clear bluish under UV radiation. Fluorescent Brightener 28 is a colorless organic compound that is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and detergents and soaps.
- This product is related structurally to Fluorescent Brightner 9 (Calcofluor White MR), which was used by Jefferies and Belcher and by Maeda and Ischida in plant tissue studies.
Specifications
Specifications
| Quantity | 5 g |
| Storage Requirements | Store at Room Temperature (15°C to 30°C) |
| Format | Powder |
| Concentration | 0.05% to 0.20% |
| For Use With (Application) | Fluorescent Brightener 28 is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and in detergents and soaps. |
| Color | Yellow |
| Melting Point | 290°C |
| CAS | 4404-43-7 |
| Molecular Formula | C40H42N12O10S2Na2 |
| Formula Weight | 960.96 |
| Show More |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.