missing translation for 'onlineSavingsMsg'
Learn More

Fluorescent Brightener 220, Technical Grade, TRC

Product Code. 30392600
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392600 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392600 Supplier Toronto Research Chemicals Supplier No. TRCF59782850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorescent Brightener 220
CAS 16470-24-9
Molecular Formula C40H40N12Na4O16S4
Quantity 50 mg
Synonym Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[[4-[bis(2-hydroxyethyl)amino]-6-[(4-sulfophenyl)amino]-1,3,5-triazin-2-yl]amino]-, sodium salt (1:4), 2,2’-Stilbenedisulfonic acid, 4,4'-bis[[4-[bis(2-hydroxyethyl)amino]-6-(p-sulfoanilino)-s-triazin-2-yl]amino]-, tetrasodium salt (7CI,8CI), Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[[4-[bis(2-hydroxyethyl)amino]-6-[(4-sulfophenyl)amino]-1,3,5-triazin-2-yl]amino]-, tetrasodium salt (9CI), BLB, Belofor OB, Blancofor BBU, Blankophor BBU, Blankophor P/BBU, Blankophor Z-JPN, C.I. 40623, C.I. Fluorescent Brightener 220, Kahablanc APU, Keyfluor White CN, Leucophor U, Photine CPK, SCRI Renaissance 2200, Sandoz CD, Tetrasodium 4,4'-bis[2-di(β-hydroxyethyl)amino-4-(4-sulfophenylamino)-s-triazin-6-ylamino]stilbine-2,2'-disulfonate
InChI Formula InChI=1S/C40H44N12O16S4.4Na/c53-19-15-51(16-20-54)39-47-35(41-27-7-11-31(12-8-27)69(57,58)59)45-37(49-39)43-29-5-3-25(33(23-29)71(63,64)65)1-2-26-4-6-30(24-34(26)72(66,67)68)44-38-46-36(48-40(50-38)52(17-21-55)18-22-56)42-28-9-13-32(14-10-28)70(60,61)62;;;;/h1-14,23-24,53-56H,15-22H2,(H,57,58,59)(H,60,61,62)(H,63,64,65)(H,66,67,68)(H2,41,43,45,47,49)(H2,42,44,46,48,50);;;;/q;4*+1/p-4
SMILES OCCN(C1=NC(NC2=CC=C(S(=O)(O[Na])=O)C=C2)=NC(NC3=C(S(=O)(O[Na])=O)C=C(/C=C/C4=CC(S(=O)(O[Na])=O)=C(NC5=NC(NC6=CC=C(S(=O)(O[Na])=O)C=C6)=NC(N(CCO)CCO)=N5)C=C4)C=C3)=N1)CCO
IUPAC Name tetrasodium;5-[[4-[bis(2-hydroxyethyl)amino]-6-(4-sulfonatoanilino)-1,3,5-triazin-2-yl]amino]-2-[2-[4-[[4-[bis(2-hydroxyethyl)amino]-6-(4-sulfonatoanilino)-1,3,5-triazin-2-yl]amino]-2-sulfonatophenyl]ethenyl]benzenesulfonate
Molecular Weight (g/mol) 1165.04
Formula Weight 1164.1159
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.