missing translation for 'onlineSavingsMsg'
Learn More

Fluorescent Brightener 134 (Technical Grade), TRC

Product Code. 30392934
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392934 500 mg
30400337 50 mg
30400338 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392934 Supplier Toronto Research Chemicals Supplier No. TRCF597825500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3426-43-5
Molecular Formula C34H28N10Na2O8S2
Molecular Weight (g/mol) 814.76
Synonym Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[[4-methoxy-6-(phenylamino)-1,3,5-triazin-2-yl]amino]-, sodium salt (1:2), 2,2’-Stilbenedisulfonic acid, 4,4'-bis[(4-anilino-6-methoxy-s-triazin-2-yl)amino]-, disodium salt (6CI,7CI,8CI), Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[[4-methoxy-6-(phenylamino)-1,3,5-triazin-2-yl]amino]-, disodium salt (9CI), Belofor KB, C.I. 40619, C.I. FBA 90, C.I. Fluorescent Brightener 134, C.I. Fluorescent Brightener 213, C.I. Fluorescent Brightener 90, C.I. Fluorescent Brightening Agent 90, CF, Disodium 4,4'-bis(2-methoxy-4-phenylamino-s-triazin-6-ylamino)stilbine-2,2'-disulfonate, Disodium 4,4'-bis[(2-anilino-4-methoxy-1,3,5-triazin-6-yl)amino]stilbene-2,2'-disulfonate, Fluorescent Brightener 213, Fluorescent Brightening Agent 90, Heliofor CSP, Heliofor RCP, Intrawite CF, Kayaphor AS, Kayaphor AS 150, Kayaphor ASC, Leucophor PC, Mikephor BS, Photine HV, Ranipal NYR, Rylux PRS, Uvitex CF, Uvitex CF 200, Uvitex CF conc. PRS, Whitex BF, Whitex BOF, Whitex HP
IUPAC Name disodium;5-[(4-anilino-6-methoxy-1,3,5-triazin-2-yl)amino]-2-[2-[4-[(4-anilino-6-methoxy-1,3,5-triazin-2-yl)amino]-2-sulfonatophenyl]ethenyl]benzenesulfonate
SMILES [Na+].[Na+].COc1nc(Nc2ccccc2)nc(Nc3ccc(C=Cc4ccc(Nc5nc(Nc6ccccc6)nc(OC)n5)cc4S(=O)(=O)[O-])c(c3)S(=O)(=O)[O-])n1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C34H30N10O8S2.2Na/c1-51-33-41-29(35-23-9-5-3-6-10-23)39-31(43-33)37-25-17-15-21(27(19-25)53(45,46)47)13-14-22-16-18-26(20-28(22)54(48,49)50)38-32-40-30(42-34(44-32)52-2)36-24-11-7-4-8-12-24;;/h3-20H,1-2H3,(H,45,46,47)(H,48,49,50)(H2,35,37,39,41,43)(H2,36,38,40,42,44);;/q;2*+1/p-2
Formula Weight 814.1328
Chemical Name or Material Fluorescent Brightener 134 (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.